Product Name
Isovaleryl-L-carnitine analytical standard
Product Number
91403
Product Brand
SIAL
CAS Number
31023-24-2
Molecular Weight
245.32
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 94.0 %
SPECIFIC ROTATION (20/D)-23.0 ± 2.0 Degrees
CONCENTRATIONC=0.1 IN WATER
WATER≤ 10.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS