Product Name
Decanoyl-L-carnitine analytical standard
Product Number
91521
Product Brand
SIAL
CAS Number
3992-45-8
Molecular Weight
315.45
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 94.0 %
SPECIFIC ROTATION (20/D)-15 ± 3 Degrees
CONCENTRATIONc=0.1 in water
WATER≤ 10.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS