Product Name
Myristoyl-L-carnitine analytical standard
Product Number
91582
Product Brand
SIAL
CAS Number
25597-07-3
Molecular Weight
371.55
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals or Flakes
PURITY (HPLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-18.0 ± 2.0 Degrees
CONCENTRATIONC=1 in Methanol
WATER≤ 10.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS