Product Name
Adipoyl-L-carnitine ≥97.0% (TLC)
Product Number
91926
Product Brand
SIAL
CAS Number
102636-83-9
Molecular Weight
289.32
Test
Specification
Assay (QNMR)*Report result
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Solid or Crystals
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-17 ± 2 Degrees
CONCENTRATIONC=0.1 in Water
WATER≤ 10 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE