Product Name
D-(+)-Melibiose analytical standard
Product Number
92413
Product Brand
SIAL
CAS Number
585-99-9
Molecular Weight
342.30
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98 %
SPECIFIC ROTATION (20/D)137.0 ± 3.0 Degrees
CONCENTRATIONC=5 IN WATER, 10H
WATER≤ 7.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS