Product Name
O-Succinyl-L-carnitine-(N-methyl-d3) lithium salt analytical standard
Product Number
92523
Product Brand
SIAL
CAS Number
Molecular Weight
264.29 (free acid basis)
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 90.0 %
IDENTIFICATION (LC-MS)Identity confirmed
Purity (LC-MS)Degree of Labelling: ≥ 99.0 %
SPECIFIC ROTATION (20/D)-19.0 ± 2.0 Degrees
CONCENTRATIONC=0.1 IN WATER
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
AAS QUALITATIVELithium confirmed
EXPIRY PERIOD36 MONTHS