Product Name
2-(Octylsulfanyl)ethan-1-ol PESTANAL®, analytical standard
Product Number
92529
Product Brand
SIAL
CAS Number
3547-33-9
Molecular Weight
190.35
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
CARBON CONTENT63.13% (Theory)
HYDROGEN CONTENT11.65% (Theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS