Product Name
Dimoxystrobin Metabolite 505M09 PESTANAL®, analytical standard
Product Number
93127
Product Brand
SIAL
CAS Number
Molecular Weight
356.37
Test
Specification
APPEARANCE (COLOR)White to Faint Brown and Faint Beige and Faint Yellow
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS