Product Name
N-Decanoylglycine analytical standard
Product Number
93204
Product Brand
SIAL
CAS Number
14305-32-9
Molecular Weight
229.32
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS