Product Name
N-Octanoylglycine analytical standard
Product Number
93529
Product Brand
SIAL
CAS Number
14246-53-8
Molecular Weight
201.26
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS