Product Name
N-Lauroylglycine analytical standard
Product Number
93586
Product Brand
SIAL
CAS Number
7596-88-5
Molecular Weight
257.37
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 90 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS