Product Name
L-Carnitine-(trimethyl-d9) inner salt analytical standard
Product Number
93689
Product Brand
SIAL
CAS Number
126827-79-0
Molecular Weight
170.25
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
IDENTIFICATION (LC-MS)Degree of Labelling: ≥99.5%
SPECIFIC ROTATION (20/D)-27.5 ± 3.0 Degrees
CONCENTRATIONc=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS