Product Name
Hexanoyl-L-carnitine-(N-methyl-d3) analytical standard
Product Number
94344
Product Brand
SIAL
CAS Number
Molecular Weight
262.36
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
IDENTIFICATION (LC-MS)Identity Confirmed
REMARKS ON LC-MSDegree of Labelling: ≥ 99.0%
PURITY (TLC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)-22 - -18 Degrees
CONCENTRATIONc=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS