Product Name
Lauroyl-L-carnitine-(N,N,N-trimethyl-d9) analytical standard
Product Number
94347
Product Brand
SIAL
CAS Number
Molecular Weight
352.56
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals or Flakes
PURITY (HPLC AREA %)≥ 95.0 %
IDENTIFICATION (LC-MS)Degree of Labelling: ≥ 99.0 %
SPECIFIC ROTATION (20/D)-20.0 ± 2.0 Degrees
CONCENTRATIONC=0.1 in Methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS