Product Name
3-Hydroxyisovaleryl-L-carnitine-(N-methyl-d3) analytical standard
Product Number
94355
Product Brand
SIAL
CAS Number
1058636-00-2
Molecular Weight
264.33
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow and White to Faint Yellow
APPEARANCE (FORM)Solid or Semi-Solid or Liquid
PURITY (HPLC AREA %)≥ 90.0 %
IDENTIFICATION (LC-MS)Identity confirmed
REMARKS ON LC-MSDegree of labelling ≥ 99.0 %
SPECIFIC ROTATION (20/D)-16.0 ± 2.0 Degrees
CONCENTRATIONc=0.1 in water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS