Product Name
4-Hydroxytamoxifen (E) and (Z) isomers (50:50), analytical standard
Product Number
94873
Product Brand
SIAL
CAS Number
68392-35-8
Molecular Weight
387.51
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.5 %
WATER≤ 5.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS