Product Name
L-Carnitine hydrochloride analytical standard
Product Number
94954
Product Brand
SIAL
CAS Number
6645-46-1
Molecular Weight
197.66
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
ASSAY (ELEMENTAL ANALYSIS)≤ 5.0 % water (calcd from elemental analysis)
SPECIFIC ROTATION (20/D)-24.0 ± 2.0 Degrees
CONCENTRATIONc=0.2 in water
CARBON CONTENT42.54 % (Theory)
HYDROGEN CONTENT8.16 % (Theory)
NITROGEN CONTENT7.09 % (Theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS