Product Name
L-Carnitine-(methyl-d3) inner salt analytical standard
Product Number
94991
Product Brand
SIAL
CAS Number
126372-70-1
Molecular Weight
164.22
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
IDENTIFICATION (LC-MS)Degree of Labelling: ≥99.5%
SPECIFIC ROTATION (20/D)-30.0 - -26.0 Degrees
CONCENTRATIONc=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS