Product Name
Sabinene hydrate analytical standard
Product Number
96573
Product Brand
SIAL
CAS Number
546-79-2
Molecular Weight
154.25
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s)
PURITY (GC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)31.5 ± 2.0 Degrees
CONCENTRATIONC=1 IN ETHANOL
MELTING POINT58 - 62 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE