Product Name
Ammonium hydroxide solution puriss., 30-33% NH3 in H2O
Product Number
05002
Product Brand
SIGALD
CAS Number
1336-21-6
Molecular Weight
35.05
Test
Specification
ASSAY30 - 33 %
RESIDUE (EVAPORATION)≤ 0.002 %
HEAVY METALS≤ 0.0003 % (AS PB)
CHLORIDE (CL)≤ 0.0002 %
SULFATE (SO4)≤ 0.002 %
IRON≤ 0.0001 %
RECOMMENDED RETEST PERIOD36 MONTHS