Product Name
Hydrochloric acid puriss., 24.5-26.0%
Product Number
07104
Product Brand
SIGALD
CAS Number
7647-01-0
Molecular Weight
36.46
Test
Specification
ASSAY24.5 - 26.0 %
SULFATED ASH≤ 0.002 %
FREE CHLORINE≤ 0.0001 %
HEAVY METALS≤ 0.0001 % (as Pb)
ARSEN≤ 0.00005 %
IRON≤ 0.0001 %
SULFATE (SO4)≤ 0.0002 %
SULFITE (SO3)≤ 0.0005 %
RECOMMENDED RETEST PERIOD42 Months