Product Name
Sulfuric acid puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Product Number
07208
Product Brand
SIGALD
CAS Number
7664-93-9
Molecular Weight
98.08
Test
Specification
ASSAY95.0 - 97.0 %
RESIDUE ON IGNITION≤ 0.0005 %
REDUCING SUBSTANCES≤ 0.0002 %
RESIDUAL SOLVENTSComplying
HEAVY METALS≤ 5 ppm (as Pb)
PROOF OF IDENTITYComplying
ARSEN≤ 0.1 ppm
CALCIUM≤ 0.001 %
CADMIUM≤ 0.0005 %
COBALT≤ 0.0005 %
CHROMIUM≤ 0.0005 %
COPPER≤ 0.0005 %
IRON≤ 2 ppm
POTASSIUM≤ 0.005 %
MAGNESIUM≤ 0.0005 %
MANGANESE≤ 0.0005 %
SODIUM≤ 0.005 %
NICKEL≤ 0.0005 %
LEAD≤ 0.0005 %
ZINC≤ 0.0005 %
CHLORIDE (CL)≤ 1 ppm
NITRATE (NO3)≤ 0.0001 %
APPEARANCE (SOLUTION)CLEAR COLORLESS
REMARKSIdentity, assay and impurities are complying to the monographs of the above mentioned pharmacopeias/codices.
RECOMMENDED RETEST PERIOD24 Months