Product Name
Ammonium bicarbonate puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%
Product Number
11213
Product Brand
SIGALD
CAS Number
1066-33-7
Molecular Weight
79.06
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) HCL 1M99.0 - 101.0 %
PHARMACOPOEA TESTSmeets analytical specification of Ph.Eur., BP, E 503
Remarks on Pharmakopoea TestIdentity comlying
non-volatile matter ≤ 100mg/kg
IRON (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 2 mg/kg
ARSENIC TRACES (MHS-AAS)≤ 1 mg/kg
MERCURY TRACES (AFS)≤ 1 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 30 mg/kg
Heavy metals (by ICP-OES)≤ 10 mg/kg
CHLORIDE (CL)≤ 10 mg/kg
RESIDUAL SOLVENTS (GLC-HS)complying
RECOMMENDED RETEST PERIOD42 MONTHS