Product Name
Sodium chloride puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
Product Number
13423
Product Brand
SIGALD
CAS Number
7647-14-5
Molecular Weight
58.44
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
ARGENTOMETR. TITRATION99.0 - 100.5 %
REMARKS ON TITRATIONcalc. to the dried substance
SOLUBILITY (COLOR)complying
SOLUBILITY (TURBIDITY)complying
SOLUBILITY (METHOD)2 G In 10 ML WATER
LOSS ON DRYING≤ 0.5 %
PHARMACOPOEA TESTSCORRESPONDS TO REQUIREMENTS
Remarks on Pharmakopoea TestAcidity or Alkalinity complying-br-Heavy metals (as Pb) ≤ 5mg/kg-br-iodide (I) test complying-br-Ferrocyanides complying-br-Nitrite (NO2) complying-br-Mg, earthalkalimetals (as Ca)-br-≤ 100mg/kg-br-Barium (Ba) complying-br-Identity complying
ALUMINIUM (ICP)≤ 0.2 mg/kg
IRON (ICP)≤ 2 mg/kg
ARSENIC TRACES (MHS-AAS)≤ 1 mg/kg
TOTAL PHOSPHORUS AS PO4 (ICP)≤ 25 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 200 mg/kg
BROMIDE (BR)≤ 50 mg/kg
RESIDUAL SOLVENTS (GLC-HS)Complying
RECOMMENDED RETEST PERIOD42 MONTHS