Product Name
Acetone puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Product Number
24201
Product Brand
SIGALD
CAS Number
67-64-1
Molecular Weight
58.08
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99 %
REMARKS ON GCrelated subst.: conforms-br-benzene ≤ 0.0002 %-br-methanol ≤ 0:05 %-br-2-propanol ≤ 0.05 %
DENSITY D20/40.790 - 0.792
SPECIFIC GRAVITY≤ 0.789
INSOLUBLE MATTERwater insoluble substances: conforms
WATER≤ 0.3 %
RESIDUE (EVAPORATION)≤ 0.002 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
APPEARANCE OF SOLUTIONconforms
ACIDITY OR ALKALINITYconforms
REDUCING SUBSTANCESconforms
RESIDUAL SOLVENTSCONFORMS
HEAVY METALS≤ 0.0001 % as Pb
REMARKSIdentity, assay and impurities are complying to the monographs of the above mentioned pharmacopeias/codices.
RECOMMENDED RETEST PERIOD60 MONTHS