Product Name
Acetone puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Product Number
32201
Product Brand
SIGALD
CAS Number
67-64-1
Molecular Weight
58.08
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 10 APHA
TITRATABLE ACID≤ 0.0015 % (AS CH3COOH)
TITRATABLE BASE≤ 0.0008 % (AS NH3)
PURITY (GC AREA %)≥ 99.5 %
REMARKS ON GCrelated subst.: conforms
benzene ≤ 0.0002 %
methanol ≤ 0.05 %
2-propanol ≤ 0.05 %
ethanol ≤ 0.01 %
DENSITY D20/40.790 - 0.792
REFRACTIVE INDEX N20/D1.3580 - 1.3600
BOILING POINT55.5 - 56.5 C
INSOLUBLE MATTERwater insoluble substances: conforms
SOLUBILITY (TURBIDITY)IN WATER: COMPLYING
WATER≤ 0.2 %
RESIDUE (EVAPORATION)≤ 0.001 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
ACIDITY OR ALKALINITYconforms
REDUCING SUBSTANCES≤ 0.0002 % (AS O)
REDUCING SUBSTANCESconforms
ALDEHYDES≤ 0.001 % (AS HCHO)
ALUMINIUM (ICP)≤ 0.5 ppm
BARIUM (ICP)≤ 0.1 ppm
BISMUTH (ICP)≤ 0.1 ppm
BORON (ICP)≤ 0.02 ppm
CALCIUM (ICP)≤ 0.5 ppm
CADMIUM (ICP)≤ 0.05 ppm
COBALT (ICP)≤ 0.01 ppm
CHROMIUM (ICP)≤ 0.02 ppm
COPPER (ICP)≤ 0.02 ppm
IRON (ICP)≤ 0.1 ppm
POTASSIUM (ICP)≤ 0.5 ppm
LITHIUM (ICP)≤ 0.1 ppm
MAGNESIUM (ICP)≤ 0.1 ppm
MANGANESE (ICP)≤ 0.01 ppm
MOLYBDENUM (ICP)≤ 0.1 ppm
SODIUM (ICP)≤ 0.5 ppm
NICKEL (ICP)≤ 0.02 ppm
LEAD (ICP)≤ 0.02 ppm
TIN (ICP)≤ 0.1 ppm
STRONTIUM (ICP)≤ 0.1 ppm
ZINC (ICP)≤ 0.05 ppm
RECOMMENDED RETEST PERIOD36 MONTHS