Product Name
Potassium formate purum p.a., ≥99.0% (NT)
Product Number
60243
Product Brand
SIGALD
CAS Number
590-29-4
Molecular Weight
84.12
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals or Chunk(s) or Powder with Chunk(s) or Crystals with Chunk(s)
TITRATION (NT) HCLO4 0.1M99.0 - 101.0 %
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 50 mg/kg
CADMIUM (ICP)≤ 50 mg/kg
COBALT (ICP)≤ 50 mg/kg
COPPER (ICP)≤ 50 mg/kg
IRON (ICP)≤ 50 mg/kg
SODIUM (ICP)≤ 5000 mg/kg
NICKEL (ICP)≤ 50 mg/kg
LEAD (ICP)≤ 50 mg/kg
ZINC (ICP)≤ 50 mg/kg
CHLORIDE (CL)≤ 50 mg/kg
SULFATE (SO4)≤ 50 mg/kg