Product Name
Sodium carbonate decahydrate puriss. p.a., ≥99.0% (T)
Product Number
71360
Product Brand
SIGALD
CAS Number
6132-02-1
Molecular Weight
286.14
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Crystals
TITRATION (T) HCL 1M99.0 - 101.0 %
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
ALUMINIUM (ICP)≤ 5 mg/kg
CALCIUM (ICP)≤ 20 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 2 mg/kg
POTASSIUM (ICP)≤ 50 mg/kg
MAGNESIUM (ICP)≤ 2 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 30 mg/kg
CHLORIDE (CL)≤ 5 mg/kg
TOTAL NITROGEN≤ 5 mg/kg
VARIOUS GUARANTEE LABEL TESTphosphate, silicate (as SiO2) ≤ 20mg/kg
RECOMMENDED RETEST PERIOD54 MONTHS