Product Name
Acacetin ≥97.0% (HPLC)
Product Number
00017
Product Brand
SIGMA
CAS Number
480-44-4
Molecular Weight
284.26
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and White to Light Brown
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE