Product Name
Atto 647N DPPE suitable for fluorescence
Product Number
05152
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Blue to Dark Blue
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)10 nmol in 20 ml DMSO
EMISSION MAXIMUM665 ± 10 nm
REMARKS ON LUMINESCENCEEXC. 643 NM IN ETHANOL
RECOMMENDED RETEST PERIOD36 MONTHS