Product Name
Atto 594 DOPE suitable for fluorescence
Product Number
05676
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Blue to Very Dark Blue
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)10 nmol in 20 ml DMSO
EMISSION MAXIMUM629 ± 10 NM
REMARKS ON LUMINESCENCEEX. 600 NM IN ETHANOL
RECOMMENDED RETEST PERIOD36 MONTHS