Product Name
Atto 594 suitable for fluorescence, ≥90.0% (HPCE)
Product Number
08637
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Faint Purple to Purple
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)Purple to Dark Purple
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 MG/ML IN DMF
EMISSION MAXIMUM627 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 601 NM IN 0.1 M PHOSPHATE PH 7.0