Product Name
Atto 594 maleimide BioReagent, suitable for fluorescence, ≥80% (coupling rate)
Product Number
08717
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Faint Purple to Purple
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Purple to Dark Purple
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 mg/ml in DMF
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM627 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 601 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD42 MONTHS