Product Name
Atto 594 NHS ester BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Product Number
08741
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Dark Blue to Violet
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Dark Red to Very Dark Red and Dark Purple to Very Dark Purple
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 MG/ML IN DMF
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM625 ± 5 nm
REMARKS ON LUMINESCENCEEX. 597 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS