Product Name
1,4-Bis(2-methylstyryl)benzene BioReagent, suitable for scintillation, ≥98% (UV)
Product Number
15090
Product Brand
SIGMA
CAS Number
13280-61-0
Molecular Weight
310.43
Test
Specification
APPEARANCE (COLOR)White to Yellow and Faint Green to Green and Yellow-Green
APPEARANCE (FORM)Powder or Crystals
MELTING POINT180 - 182 C
SOLUBILITY (COLOR)Colorless to Light Yellow and Colorless to Light Green-Yellow
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)0.5G IN 10ML DIOXANE, HOT
CARBON CONTENT92.86 % (THEORY)
HYDROGEN CONTENT7.14 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
WAVELENGTH (1) (UV)350 - 360 nm
ASSAY (1) (UV)≥ 98 %
SOLVENT (UV)Dioxane
REMARKSsuitable as laser dye
RECOMMENDED RETEST PERIOD42 MONTHS