Product Name
5(6)-Carboxyfluorescein diacetate BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Product Number
21879
Product Brand
SIGMA
CAS Number
124387-19-5
Molecular Weight
460.39
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 90.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM517 ± 5 nm