Product Name
5(6)-Carboxynaphthofluorescein BioReagent, suitable for fluorescence, ≥90% (HPLC)
Product Number
21932
Product Brand
SIGMA
CAS Number
128724-35-6
Molecular Weight
476.43
Test
Specification
APPEARANCE (COLOR)DEEP BLUE TO DEEP VIOLET
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 90.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM672 ± 5 nm