Product Name
Quinine hydrochloride dihydrate BioReagent, suitable for fluorescence, ≥98.0% (NT)
Product Number
22630
Product Brand
SIGMA
CAS Number
6119-47-7
Molecular Weight
396.91
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M98.0 - 102.0 %
PURITY (HPLC AREA %)≥ 90 %
MINOR COMPONENT 1DIHYDROQUININE HYDROCHLORIDE
HPLC MINOR COMPONENT (AREA %)≤ 10 %
SPECIFIC ROTATION (20/D)-235.0 - -225.0 Degrees
CONCENTRATIONC=2 IN HYDROCHLORIC ACID 0.1M
LOSS ON DRYING6 - 10 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM440.0 - 456.0 nm