Product Name
Atto 425-Biotin BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Product Number
28616
Product Brand
SIGMA
CAS Number
Molecular Weight
711.91
Test
Specification
APPEARANCE (COLOR)White to Light Green
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 95.0 %
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM481 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 437 nm in 0.1 M phosphate buffer pH 7.0