Product Name
Dibromobimane BioReagent, suitable for fluorescence, ≥95.0% (CHN)
Product Number
34025
Product Brand
SIGMA
CAS Number
68654-25-1
Molecular Weight
350.01
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals
ASSAY (ELEMENTAL ANALYSIS)≥ 95 % (CHN)
CARBON CONTENT34.32 % (THEORY)
HYDROGEN CONTENT2.88 % (THEORY)
NITROGEN CONTENT8.00 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM477 ± 5 nm
REMARKS ON LUMINESCENCEExc.: 393 nm in 0.1 M tris, pH 7.0; red. glutathione
SUITABILITY ASSAYsuitable for the fluorogenic derivatisation of thiols