Product Name
Atto 590 maleimide BioReagent, suitable for fluorescence
Product Number
39887
Product Brand
SIGMA
CAS Number
Molecular Weight
813.29
Test
Specification
APPEARANCE (COLOR)Red or Purple
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMSO
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM622 ± 5 nm
REMARKS ON LUMINESCENCEEX. 595 NM IN 0.1 M PHOSPHATE PH 7.0; 2-MERCAPTOETHANOL