Product Name
Atto 488 suitable for fluorescence, ≥90% (HPCE)
Product Number
41051
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Red to Dark Red
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMSO
EMISSION MAXIMUM521 ± 5 nm
REMARKS ON LUMINESCENCEExc. 504 nm in 0.1 M phosphate pH 7.0
RECOMMENDED RETEST PERIOD72 MONTHS