Product Name
Atto 610 maleimide BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Product Number
41061
Product Brand
SIGMA
CAS Number
Molecular Weight
613.10
Test
Specification
APPEARANCE (COLOR)VIOLET TO DEEP BLUE
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)VIOLET TO DEEP BLUE
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 MG/ML IN DMSO
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
DEGREE OF COUPLING≥ 90.0 %
EMISSION MAXIMUM637.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEXC. 610 NM IN 0.1 M PHOSPHATE PH 7.0