Product Name
Atto 635 NHS ester BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Product Number
43429
Product Brand
SIGMA
CAS Number
Molecular Weight
628.11
Test
Specification
APPEARANCE (COLOR)Dark Blue to Very Dark Blue
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)report result
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 mg/ml in DMF
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM655 ± 5 nm
REMARKS ON LUMINESCENCEEX. 624 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD28 MONTHS