Product Name
Fluorescein isothiocyanate–dextran average mol wt 70,000, (FITC:Glucose = 1:250)
Product Number
46945
Product Brand
SIGMA
CAS Number
60842-46-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Yellow to Very Dark Yellow and Light Orange to Very Dark Orange
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)Orange to Very Dark Orange
SOLUBILITY (TURBIDITY)Clear to Turbid
SOLUBILITY (METHOD)50MG/ML WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM513 - 523 nm
REMARKS ON LUMINESCENCEEXC.: 492 NM IN 0.1 M PHOSPHATE PH 8.0