Product Name
Fluorescein isothiocyanate – Dextran 500000-Conjugate (FITC:Glucose = 1:100)
Product Number
46947
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Yellow to Dark Yellow
APPEARANCE (FORM)Powder or Crystals
SOLUBILITY (COLOR)Yellow and Orange and Yellow-Orange
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)25 MG/ML WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
MISCELLANEOUS TESTS0.003 - 0.020 MOL FITC/MOL OF GLUCOSE