Product Name
4-Fluoro-7-nitrobenzofurazan BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Product Number
47140
Product Brand
SIGMA
CAS Number
29270-56-2
Molecular Weight
183.10
Test
Specification
APPEARANCE (COLOR)Faint Yellow to Yellow
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98.0 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.1G IN 10ML CHLOROFORM
CARBON CONTENT39.6 - 0.0 %
HYDROGEN CONTENT1.1 - 0.0 %
NITROGEN CONTENT22.9 - 0.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
FLUORINE 19 NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
RECOMMENDED RETEST PERIOD42 MONTHS