Product Name
Atto Rho101 NHS ester
Product Number
50492
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)REPORT RESULT
APPEARANCE (FORM)REPORT RESULT
DEGREE OF COUPLING≥ 80.0 %
EMISSION MAXIMUM610 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 586 NM IN 0.1 M PHOSPHATE PH 7.0