Product Name
Atto 465 BioReagent, suitable for fluorescence
Product Number
50712
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Dark Red to Very Dark Red and Red-Brown and Very Dark Red-Brown
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 95.0 %
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)2 mg/ml in Methanol
EMISSION MAXIMUM504 ± 5 nm
REMARKS ON LUMINESCENCEEXC. 454 NM IN 0.1 M PHOSPHATE PH 7.0