Product Name
Atto 740-Biotin suitable for fluorescence, ≥90.0% (HPCE)
Product Number
50842
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)REPORT RESULT
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 90.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
EMISSION MAXIMUM764 ± 5 nm
REMARKS ON LUMINESCENCEEX. 740 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS